Anti-BCL2L14 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7585R
10465-198EA 796.88 CAD
10465-198
Anti-BCL2L14 Rabbit Polyclonal Antibody
Antibodies
Plays a role in apoptosis.

Recommended Dilutions: IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: BCL2L14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR