Anti-A3GALNT, A3GALT1, GTB, NAGAT Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7589R
10465-294EA 796.89 CAD
10465-294
Anti-A3GALNT, A3GALT1, GTB, NAGAT Rabbit Polyclonal Antibody
Antibodies
This gene encodes proteins related to the first discovered blood group system, ABO. Which allele is present in an individual determines the blood group. The 'O' blood group is caused by a deletion of guanine-258 near the N-terminus of the protein which results in a frameshift and translation of an almost entirely different protein. Individuals with the A, B, and AB alleles express glycosyltransferase activities that convert the H antigen into the A or B antigen. Other minor alleles have been found for this gene.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: A3GALNT, A3GALT1, GTB, NAGAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR