Anti-IKIP Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7600R
10465-504EA 796.88 CAD
10465-504
Anti-IKIP Rabbit Polyclonal Antibody
Antibodies
Target of p53/TP53 with pro-apoptotic function.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: IKIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR