Anti-TIAF1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7602R
10464-912EA 796.88 CAD
10464-912
Anti-TIAF1 Rabbit Polyclonal Antibody
Antibodies
TIAF-1 (TGF Beta 1-induced antiapoptotic factor 1), also known as MYO18A, MAJN or MYSPDZ, is a TGF Beta 1-induced anti-apoptotic factor that localizes to the nucleus and functions to inhibit the cytotoxic effects of TNF? TRADD and FADD. Two isoforms of mouse TIAF-1 exist and are designated MysPDZ-Alpha and MysPDZ-Beta. MysPDZ-Alpha consists of a KE-rich region, an N-terminal PDZ domain and a prevalent myosin homologous head region, while the MysPDZ-Beta isoform lacks the KE-rich region and PDZ domain. MysPDZ-Alpha is present in most tissues and is known to co-localize with the ER-Golgi complex and with membrane ruffles and filopodia. MysPDZ-Beta is expressed specifically in hematopoietic tissues and cell lines and shows dispersed localization in the cytoplasm.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: TIAF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR