Anti-Cipar1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7603R
10465-560EA 796.88 CAD
10465-560
Anti-Cipar1 Rabbit Polyclonal Antibody
Antibodies
Cipar1 may be involved in a survival program enabling certain prostatic cells to resist apoptosis. It is expressed in prostate and detected in other organs at low levels, these include the heart and various tissues of the urogenital tract; not detected in mammary gland.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: Cipar1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR