Anti-KIAA0101 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7627R
10466-068EA 796.88 CAD
10466-068
Anti-KIAA0101 Rabbit Polyclonal Antibody
Antibodies
NS5ATP9/PAF/p15(PAF) is involved in protection of cells from UV-induced cell death and is also a Hepatitis C NS5A up-regulation gene which may play a role in the pathogenesis of HCV-associated hepatocellular carcinoma.

Recommended Dilutions: IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: KIAA0101
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR