Anti-GHITM Rabbit Polyclonal Antibody

BS-7629R
10466-116EA 397.94 CAD
10466-116
Anti-GHITM Rabbit Polyclonal Antibody
Required for the mitochondrial tubular network and cristae organization. Involved in apoptotic release of cytochrome c.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: MICS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR