Anti-AGTRAP Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7642R
10466-356EA 796.88 CAD
10466-356
Anti-AGTRAP Rabbit Polyclonal Antibody
Antibodies
AGTRAP (Angiotensin II receptor-associated protein) interacts with the angiotensin II type 1 receptor and negatively regulates type 1 angiotensin II receptor-mediated signaling, by regulating receptor internalisation as well as mechanisms of receptor desensitization such as phosphorylation.

Recommended Dilutions: IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: AGTRAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR