Anti-PDCD2L Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7666R
10466-838EA 796.88 CAD
10466-838
Anti-PDCD2L Rabbit Polyclonal Antibody
Antibodies
MGC13096, also known as PDCD2L, might play a role in apoptosis given evidence that over expression attenutates TNF alpha release in Daudi cells anad restrained proliferation of HEK293T cells.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: PDCD2L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR