Anti-FKBPL Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7681R
10467-180EA 796.88 CAD
10467-180
Anti-FKBPL Rabbit Polyclonal Antibody
Antibodies
May be involved in response to X-ray. Regulates p21 protein stability by binding to Hsp90 and p21.Tissue specificity: Ubiquitously expressed with higher levels in testis.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: WISP39
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR