Anti-TP53TG3 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7701R
10467-792EA 796.88 CAD
10467-792
Anti-TP53TG3 Rabbit Polyclonal Antibody
Antibodies
May play a significant role in TP53/p53-mediating signaling pathway.Tissue specificity:Strongly expressed in testis. Weakly expressed in heart, placenta and skeletal muscle.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: TP53TG3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR