Anti-TP53TG5 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7702R
10467-818EA 796.88 CAD
10467-818
Anti-TP53TG5 Rabbit Polyclonal Antibody
Antibodies
TP53TG5 may play a significant role in p53/TP53-mediating signaling pathway. It is highly expressed in heart, brain and small intestin. Less abundant in skeletal muscle, spleen, prostate, ovary and colon. A smaller transcript is expressed specifically in the testis.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: TP53TG5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR