Anti-CENPP Rabbit Polyclonal Antibody

BS-7710R
10467-906EA 397.94 CAD
10467-906
Anti-CENPP Rabbit Polyclonal Antibody
CENPP is a component of the CENPA-CAD (nucleosome distal) complex, a complex recruited to centromeres which is involved in assembly of kinetochore proteins, mitotic progression and chromosome segregation.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CENPP
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR