Anti-NCAPD3 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7734R
10468-474EA 796.86 CAD
10468-474
Anti-NCAPD3 Rabbit Polyclonal Antibody
Antibodies
Condensin complexes I and II play essential roles in mitotic chromosome assembly and segregation. Condensins contain 2 invariant structural maintenance of chromosome (SMC) subunits, SMC2 and SMC4. hCAP-D3 is a regulatory non-SMC subunit of the condensin II complex.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: NCAPD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR