Anti-KLHL9 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7753R
10468-946EA 796.88 CAD
10468-946
Anti-KLHL9 Rabbit Polyclonal Antibody
Antibodies
KLHL9 is the substrate-specific adapter for a CUL3-based E3 ubiquitin-protein ligase complex. Within this complex, KLHL9 controls the dynamic behavior of AURKB on mitotic chromosomes and thereby coordinates faithful mitotic progression and completion of cytokinesis.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: KLHL9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR