Anti-SEPT10 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7770R
10469-410EA 796.88 CAD
10469-410
Anti-SEPT10 Rabbit Polyclonal Antibody
Antibodies
The SEPT10 gene encodes a member of the septin family of cytoskeletal proteins which have GTPase activity. This protein localizes to the cytoplasm and nucleus and displays GTP-binding and GTPase activity. Alternate splicing results in two transcript variants encoding different isoforms.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: SEPT10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR