Anti-CCNB1IP1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7784R
10469-642EA 796.88 CAD
10469-642
Anti-CCNB1IP1 Rabbit Polyclonal Antibody
Antibodies
E3 ubiquitin-protein ligase. Modulates cyclin B levels and participates in the regulation of cell cycle progression through the G2 phase. Overexpression causes delayed entry into mitosis.Tissue specificity: Highly expressed in heart. Detected at intermediate levels in liver and kidney, and at low levels in placenta, brain and lung.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCNB1IP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR