Anti-CEP152 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7787R
10469-274EA 796.88 CAD
10469-274
Anti-CEP152 Rabbit Polyclonal Antibody
Antibodies
Defects in CEP152 are the cause of microcephaly primary type 4 (MCPH4). A disease defined as a head circumference more than 3 standard deviations below the age-related mean. Brain weight is markedly reduced and the cerebral cortex is disproportionately small. Despite this marked reduction in size, the gyral pattern is relatively well preserved, with no major abnormality in cortical architecture. Affected individuals are mentally retarded. Primary microcephaly is further defined by the absence of other syndromic features or significant neurological deficits due to degenerative brain disorder.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CEP152
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR