Anti-DSN1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7796R
10469-962EA 796.88 CAD
10469-962
Anti-DSN1 Rabbit Polyclonal Antibody
Antibodies
This gene encodes a kinetochore protein that functions aspart of the minichromosome instability-12 centromere complex. Theencoded protein is required for proper kinetochore assembly andprogression through the cell cycle. Alternative splicing results inmultiple transcript variants. [provided by RefSeq, Feb 2009].

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: DSN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR