Anti-KLHDC5 Rabbit Polyclonal Antibody

BS-7806R
10470-084EA 397.94 CAD
10470-084
Anti-KLHDC5 Rabbit Polyclonal Antibody
KLHDC5 contains 1 BTB (POZ) domain and 6 Kelch repeats. It is phosphorylated upon DNA damage, possibly by ATM or ATR. The exact function of KLHDC5 is currently unknown.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: KLHDC5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR