Anti-MPP9 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7812R
10470-294EA 796.62 CAD
10470-294
Anti-MPP9 Rabbit Polyclonal Antibody
Antibodies
Biological process: M phase of mitotic cell cycle. There are two isoforms.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: MPP9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR