Anti-NEK8 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7815R
10470-150EA 796.88 CAD
10470-150
Anti-NEK8 Rabbit Polyclonal Antibody
Antibodies
Required for renal tubular integrity.Tissue specificity: Highest expression in thyroid, adrenal gland and skin. Low levels in spleen, colon and uterus. Overexpressed in breast tumors, with highest expression in infiltrating ductal carcinomas and moderate levels in mucinous adenocarcinoma.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: NEK8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR