Anti-SPECC1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7816R
10470-342EA 796.88 CAD
10470-342
Anti-SPECC1 Rabbit Polyclonal Antibody
Antibodies
NSP type 5 alpha 3 alpha is a novel structure protein belonging to the cytospin-A family, that may play a role in nuclear function during cell division. It is also highly expressed in certain tumor cell lines, suggesting that it may serve as a good tumor marker. It was was cloned as part of a chromosomal translocation involving PDGFRB in a case of juvenile myelomonocytic leukemia.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: SPECC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR