Anti-NUBP1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7817R
10470-366EA 796.88 CAD
10470-366
Anti-NUBP1 Rabbit Polyclonal Antibody
Antibodies
Implicated in the regulation of centrosome duplication (By similarity). Component of the cytosolic iron-sulfur (Fe/S) protein assembly machinery. Required for maturation of extramitochondrial Fe/S proteins. May bind and transfer 2 labile 4Fe-4S clusters to target apoproteins.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: NUBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR