Anti-SKA2 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7847R
10470-542EA 796.88 CAD
10470-542
Anti-SKA2 Rabbit Polyclonal Antibody
Antibodies
Ska2 (spindle and kinetochore associated complex subunit 2), also known as FAM33A, is a 121 amino acid component of the Ska1 complex, a microtubule-binding subcomplex of the outer kinetochore that is critical for proper chromosome segregation. The Ska1 complex is a component of the kinetochore-microtubule interface and directly associates with microtubules as oligomeric assemblies. Localized to the outer kinetochore and spindle microtubules during cell proliferation, Ska2 is essential for spindle checkpoint silencing and exit from mitosis. Downregulation of Ska2 leads to delayed recruitment of MAD2, a component of the mitotic spindle checkpoint, to several kinetochores resulting in occasional loss of individual chromosomes from the metaphase plate. Ska2 is encoded by a gene located on human chromosome 17, which comprises over 2.5% of the human genome and encodes over 1,200 genes.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: SKA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR