Anti-ZWINT Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7852R
10471-182EA 796.88 CAD
10471-182
Anti-ZWINT Rabbit Polyclonal Antibody
Antibodies
ZWINT is a component of the MIS12 complex, which is required for kinetochore function and spindle checkpoint activity. ZWINT interacts with ZW10 and targets it to the kinetochore at prometaphase. ZWINT is detectable on the kinetochore till late anaphase. Inhibition of ZWINT expression results in chromosome missegregation, spindle checkpoint failure and, ultimately, cell death upon cytokinesis.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZWINT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR