Anti-ALOXE3 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7856R
10471-310EA 796.88 CAD
10471-310
Anti-ALOXE3 Rabbit Polyclonal Antibody
Antibodies
Introduces molecular oxygen into polyunsaturated fatty acids. Exact substrate is not known.Tissue specificity:Predominantly expressed in skin.Involvement in diseaseDefects in ALOXE3 are a cause of non-bullous congenital ichthyosiform erythroderma (NCIE). NCIE is a non-bullous ichthyosis, a skin disorder characterized by abnormal cornification of the epidermis. Most affected individuals are born with a tight, shiny, translucent covering called collodion membrane. The collodion membrane subsequently evolves into generalized scaling and intense redness of the skin. Clinical features are milder than in lamellar ichthyoses and demonstrate a greater variability in the intensity of erythema, size and type of scales. In contrast to lamellar ichthyoses, scales are usually white, fine and powdery, and palms and soles are severely affected. Patients suffer from palmoplantar keratoderma, often with painful fissures, digital contractures, and loss of pulp volume.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: ALOXE3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR