Anti-S100 A2 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7873R
10471-686EA 796.89 CAD
10471-686
Anti-S100 A2 Rabbit Polyclonal Antibody
Antibodies
The protein encoded by this gene is a member of the S100family of proteins containing 2 EF-hand calcium-binding motifs.S100 proteins are localized in the cytoplasm and/or nucleus of awide range of cells, and involved in the regulation of a number ofcellular processes such as cell cycle progression anddifferentiation. S100 genes include at least 13 members which arelocated as a cluster on chromosome 1q21. This protein may have atumor suppressor function. Chromosomal rearrangements and alteredexpression of this gene have been implicated in breast cancer.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: S100 A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR