Anti-SPAG8 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7876R
10471-758EA 796.88 CAD
10471-758
Anti-SPAG8 Rabbit Polyclonal Antibody
Antibodies
May play a role in fertility and microtubule formation through interaction with RANBP9.

Recommended Dilutions: IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: SPAG8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR