Anti-SPDYA Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7901R
10472-310EA 796.61 CAD
10472-310
Anti-SPDYA Rabbit Polyclonal Antibody
Antibodies
Regulates the G1/S phase transition of the cell cycle by binding and activating CDK1, CDK2 and CDKN1B/KIP1. Can activate CDK2 without promoting CDK2 phosphorylation. Mediates cell survival during the DNA damage process through activation of CDK2.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: SPDYA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR