Anti-CK II beta Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7909R
10472-512EA 796.86 CAD
10472-512
Anti-CK II beta Rabbit Polyclonal Antibody
Antibodies
Plays a complex role in regulating the basal catalytic activity of the alpha subunit (By similarity). Participates in Wnt signaling.

Recommended Dilutions: IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CK II beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR