Anti-TLK2 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7925R
10472-896EA 796.88 CAD
10472-896
Anti-TLK2 Rabbit Polyclonal Antibody
Antibodies
Rapidly and transiently inhibited by phosphorylation following the generation of DNA double-stranded breaks during S-phase. This is cell cycle checkpoint and ATM-pathway dependent and appears to regulate processes involved in chromatin assembly.Tissue specificity: Widely expressed. Present in fetal placenta, liver, kidney, pancreas, heart and skeletal muscle. Also found in adult cell lines.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: TLK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR