Anti-ANKRD28 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7926R
10472-920EA 796.88 CAD
10472-920
Anti-ANKRD28 Rabbit Polyclonal Antibody
Antibodies
ANKRD28 selectively inhibits the phosphatase activity of PPP1C and targets PPP1C to modulate HNRPK phosphorylation. It contains twenty seven ANK repeats. There are two named isoforms.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: ANKRD28
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR