Anti-PPP2R1A Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7931R
10473-064EA 796.88 CAD
10473-064
Anti-PPP2R1A Rabbit Polyclonal Antibody
Antibodies
Protein phosphatase 2A (PP2A) is the most abundant phosphatase specific for serine and threonine phosphorylation in mammalian cells. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The constant regulatory subunit alpha (PPP2R1A) serves as a scaffolding molecule to coordinate the assembly of the catalytic subunit and a variable regulatory beta subunit.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: PPP2R1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR