Anti-ABHD13 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7943R
10473-112EA 796.88 CAD
10473-112
Anti-ABHD13 Rabbit Polyclonal Antibody
Antibodies
Ubiquitously expressed.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: ABHD13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR