Anti-ABHD15 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7946R
10473-416EA 796.88 CAD
10473-416
Anti-ABHD15 Rabbit Polyclonal Antibody
Antibodies
ABHD15

Recommended Dilutions: IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: ABHD15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR