Anti-ABHD3 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7947R
10473-440EA 796.88 CAD
10473-440
Anti-ABHD3 Rabbit Polyclonal Antibody
Antibodies
Abhydrolase domain containing 3 (ABHD3) belongs to the AB hydrolase superfamily. It has a central core predicted to form alpha/beta hydrolase folds and has a ser-ala-his catalytic triad characteristic of peroxidases.

Recommended Dilutions: IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: ABHD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR