Anti-APM2 Rabbit Polyclonal Antibody

Supplier: Bioss
BS-7965R
10473-906EA 677.18 CAD
10473-906
Anti-APM2 Rabbit Polyclonal Antibody
APM2 gene is exclusively expressed in adipose tissue. Its function is currently unknown. APM2 has been shown to be associated with cisplatin resistance in cell lines. It is not found in cisplatin sensitive cell lines, implicating APM2 in the development of cisplatin resistance in tumor cells.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: APM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR