Anti-APOL3 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7967R
10473-682EA 796.88 CAD
10473-682
Anti-APOL3 Rabbit Polyclonal Antibody
Antibodies
APOL3 may be involved in cytoplasmic lipid transport or allow the binding of lipids to organelles. It is widely expressed; the highest levels are in prostate, lung and placenta; also detected in kidney, bone marrow, spleen, thymus, spinal cord, adrenal gland, salivary gland, trachea and mammary gland; levels are low in brain, heart, fetal liver, pancreas and testis. There are three named isoforms.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: APOL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR