Anti-SRRT Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7970R
10473-754EA 796.88 CAD
10473-754
Anti-SRRT Rabbit Polyclonal Antibody
Antibodies
ARS2(Arsenite resistance protein 2) confers arsenite resistance. The protein belongs to the ARS2 family. Arsenic is a human carcinogen whose mechanism of action is unknown. The arsenite acts as a comutagen by interfering with DNA repair. Two genes, ASR1(Arsenite resistance protein 1) and ASR2 (Arsenite resistance protein 2), confer arsenite resistance to arsenite-sensitive cells. ASR1 shows almost complete homology with the rat fau gene, a tumor suppressor gene which contains a ubiquitin like region fused to S30 ribosomal protein. Arsenite inhibits ubiquitin dependent proteolysis. The tumor suppressor fau gene product or some other aspect of the ubiquitin system may be a target for arsenic toxicity and that disruption of the ubiquitin system may contribute to the genotoxicity and carcinogenicity of arsenite.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: SRRT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR