Anti-ATE1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7973R
10473-994EA 796.88 CAD
10473-994
Anti-ATE1 Rabbit Polyclonal Antibody
Antibodies
ATE1, also known as arginyltransferase 1, is an enzyme that is involved in the post-translational conjugation of arginine to the N-terminal aspartate or glutamate of a protein. This arginylation is required for degradation of the protein via the ubiquitin pathway. There are two named isoforms.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: ATE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR