Anti-CA7 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7984R
10474-266EA 796.88 CAD
10474-266
Anti-CA7 Rabbit Polyclonal Antibody
Antibodies
Carbonic anhydrases are a large family of zincmetalloenzymes that catalyze the reversible hydration of carbondioxide. They participate in a variety of biological processes,including respiration, calcification, acid-base balance, boneresorption, and the formation of aqueous humor, cerebrospinalfluid, saliva, and gastric acid. They show extensive diversity intissue distribution and in their subcellular localization. Thecytosolic protein encoded by this gene is predominantly expressedin the salivary glands. Alternative splicing in the coding regionresults in multiple transcript variants encoding differentisoforms. [provided by RefSeq, Jul 2008].

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CA7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR