Anti-CCBE1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7985R
10474-322EA 796.88 CAD
10474-322
Anti-CCBE1 Rabbit Polyclonal Antibody
Antibodies
This gene is thought to function in extracellular matrixremodeling and migration. It is predominantly expressed in theovary, but down regulated in ovarian cancer cell lines and primarycarcinomas, suggesting its role as a tumour suppressor. Mutationsin this gene have been associated with Hennekamlymphangiectasia-lymphedema syndrome, a generalized lymphaticdysplasia in humans. [provided by RefSeq, Mar 2010].

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCBE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR