Anti-CCDC102B Rabbit Polyclonal Antibody

BS-7986R
10474-346EA 397.94 CAD
10474-346
Anti-CCDC102B Rabbit Polyclonal Antibody
CCDC102B

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCDC102B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR