Anti-CCDC16 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7987R
10474-370EA 796.88 CAD
10474-370
Anti-CCDC16 Rabbit Polyclonal Antibody
Antibodies
This gene encodes for a coiled coil domain containing 16 protein and is located on chromosome 17. The protein may act as a regulator of the cell cycle in embryos by participating in control of M phase.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCDC16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR