Anti-CDAN1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7994R
10474-554EA 796.88 CAD
10474-554
Anti-CDAN1 Rabbit Polyclonal Antibody
Antibodies
This gene encodes a protein that appears to play a role in nuclear envelope integrity, possibly related to microtubule attachments. Mutations in this gene cause congenital dyserythropoietic anemia type I, a disease resulting in morphological and functional abnormalities of erythropoiesis.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CDAN1
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR