Anti-COBLL1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8031R
10475-194EA 796.88 CAD
10475-194
Anti-COBLL1 Rabbit Polyclonal Antibody
Antibodies
The function of COBLL1 is not known but there are five named isoforms produced by alternative splicing.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: COBLL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR