Anti-GABARAPL1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8035R
10475-298EA 796.88 CAD
10475-298
Anti-GABARAPL1 Rabbit Polyclonal Antibody
Antibodies
GABARAPL1 belongs to the MAP1 LC3 family. It increases cell surface expression of kappa type opioid receptor through facilitating anterograde intracellular trafficking of the receptor.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: GABARAPL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR