Anti-COMMD4 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8037R
10475-346EA 796.88 CAD
10475-346
Anti-COMMD4 Rabbit Polyclonal Antibody
Antibodies
COMMD is a new family of proteins with homology to MURR1, a multifunctional protein that inhibits NFkB. These proteins form multimeric complexes and were identified in a biochemical screen for MURR1-associated factors. The family is defined by the presence of a conserved and unique motif termed the COMM (copper metabolism gene MURR1) domain, which functions as an interface for protein-protein interactions. Down-regulates activation of NF-kappa-B.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: COMMD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR