Anti-KLHL41 Rabbit Polyclonal Antibody

Supplier: Bioss
BS-8044R
10475-500EA 697.49 CAD
10475-500
Anti-KLHL41 Rabbit Polyclonal Antibody
Sarcosin contains 1 BTB (POZ) domain and is required for pseudopod elongation in transformed cells. Sarcosin mRNA is up-regulated by less than two folds in the heart in human patients with HCM.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: KLHL41
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR